ENA313838976 Enamine

4-methyl-2-(pyrrolidin-1-yl)pentan-1-amine hydrochloride

  • MDL number MFCD18380615
  • Cas Number : 1269152-35-3
  • Molecular Weight : 206.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72439
Availability details for
Quantity
  • Availability
  • Add to Cart