ENA313838985 Enamine

(3S)-3-amino-3-(5-methylfuran-2-yl)propanoic acid hydrochloride

  • MDL number MFCD18380625
  • Cas Number : 1305712-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72565
Availability details for
Quantity
  • Availability
  • Add to Cart