ENA313838990 Enamine

3-[1-(methylamino)ethyl]phenol hydrochloride

  • MDL number MFCD18380629
  • Cas Number : 1269152-23-9
  • Molecular Weight : 187.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72619
Availability details for
Quantity
  • Availability
  • Add to Cart