ENA313838991 Enamine

(2S)-2-amino-1-(piperidin-1-yl)propan-1-one hydrochloride

  • MDL number MFCD18380631
  • Cas Number : 916762-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72632
Availability details for
Quantity
  • Availability
  • Add to Cart