ENA313838998 Enamine

4-[(5-methyl-1,2,4-oxadiazol-3-yl)methoxy]aniline hydrochloride

  • MDL number MFCD18380639
  • Cas Number : 1269151-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-72684

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72684
Availability details for
Quantity
  • Availability
  • Add to Cart