ENA313839000 Enamine

3-(propan-2-yl)-5-(trichloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD18380641
  • Cas Number : 1199-49-1
  • Molecular Weight : 229.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72691
Availability details for
Quantity
  • Availability
  • Add to Cart