ENA313839001 Enamine

2-[5-(pyridin-2-yl)-1,2,4-oxadiazol-3-yl]ethan-1-amine hydrochloride

  • MDL number MFCD18380642
  • Cas Number : 1269152-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72701
Availability details for
Quantity
  • Availability
  • Add to Cart