ENA313839002 Enamine

N-amino-2,5-difluorobenzene-1-carboximidamide

  • MDL number MFCD18380644
  • Cas Number : 1029623-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72713
Availability details for
Quantity
  • Availability
  • Add to Cart