ENA313839005 Enamine

4-(aminomethyl)-N-methyl-1,3-thiazol-2-amine dihydrochloride

  • MDL number MFCD18380647
  • Cas Number : 1269152-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72731
Availability details for
Quantity
  • Availability
  • Add to Cart