ENA313839011 Enamine

2-chloro-N-{2-[(2-cyano-2-methylethyl)sulfanyl]phenyl}acetamide

  • MDL number MFCD18380654
  • Cas Number : 1269152-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72787

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72787
Availability details for
Quantity
  • Availability
  • Add to Cart