ENA313839016 Enamine

ethyl 2-(cyclopropylamino)-4-methyl-1,3-thiazole-5-carboxylate hydrochloride

  • MDL number MFCD18380660
  • Cas Number : 1269152-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72814

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72814
Availability details for
Quantity
  • Availability
  • Add to Cart