ENA313839018 Enamine

ethyl 4-methyl-2-(methylamino)-1,3-thiazole-5-carboxylate hydrochloride

  • MDL number MFCD18380662
  • Cas Number : 1269152-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218...220
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72816

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72816
Availability details for
Quantity
  • Availability
  • Add to Cart