ENA313839020 Enamine

1-{[3-(aminomethyl)phenyl]methyl}pyrrolidin-2-one hydrochloride

  • MDL number MFCD18380664
  • Cas Number : 1269152-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72853
Availability details for
Quantity
  • Availability
  • Add to Cart