ENA313839021 Enamine

2-(2-tert-butylphenoxy)ethan-1-amine hydrochloride

  • MDL number MFCD18380665
  • Cas Number : 1269152-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72856
Availability details for
Quantity
  • Availability
  • Add to Cart