ENA313839027 Enamine

2-(1H-1,3-benzodiazol-1-yl)-2-phenylacetic acid hydrochloride

  • MDL number MFCD18380671
  • Cas Number : 1269151-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72881

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72881
Availability details for
Quantity
  • Availability
  • Add to Cart