ENA313839028 Enamine

3-(methanesulfonylmethyl)-5-(trichloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD18380672
  • Cas Number : 1269152-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72887
Availability details for
Quantity
  • Availability
  • Add to Cart