ENA313839030 Enamine

4-amino-1-methylpyrrolidin-2-one hydrochloride

  • MDL number MFCD09864324
  • Cas Number : 1228838-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72905

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-72905
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart