ENA313839032 Enamine

2-(6-fluoro-1-methyl-1H-1,3-benzodiazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD18380676
  • Cas Number : 1269151-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72910
Availability details for
Quantity
  • Availability
  • Add to Cart