ENA313839033 Enamine

2-(4-fluoro-1-methyl-1H-1,3-benzodiazol-2-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD18380677
  • Cas Number : 1269152-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72911
Availability details for
Quantity
  • Availability
  • Add to Cart