ENA313839036 Enamine

3-(4-bromo-2-fluorophenyl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD18380681
  • Cas Number : 1016771-38-2
  • Molecular Weight : 257.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72941
Availability details for
Quantity
  • Availability
  • Add to Cart