ENA313839039 Enamine

(4-fluorophenyl)(quinolin-3-yl)methanamine dihydrochloride

  • MDL number MFCD18380685
  • Cas Number : 1269152-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72952

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72952
Availability details for
Quantity
  • Availability
  • Add to Cart