ENA313839043 Enamine

3-(1H-1,2,4-triazol-1-ylmethyl)piperidine dihydrochloride

  • MDL number MFCD18380689
  • Cas Number : 1269152-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73001

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73001
Availability details for
Quantity
  • Availability
  • Add to Cart