ENA313839047 Enamine

ethyl 5-amino-1-[2-(methanesulfonyloxy)ethyl]-1H-pyrazole-4-carboxylate

  • MDL number MFCD18380693
  • Cas Number : 126352-83-8
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73018
Availability details for
Quantity
  • Availability
  • Add to Cart