ENA313839062 Enamine

tert-butyl N-{4-[(5-methyl-1,3,4-oxadiazol-2-yl)methoxy]phenyl}carbamate

  • MDL number MFCD18380709
  • Cas Number : 1269152-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73122
Availability details for
Quantity
  • Availability
  • Add to Cart