ENA313839066 Enamine

N-methyl-N-(piperidin-4-yl)ethane-1-sulfonamide hydrochloride

  • MDL number MFCD18380714
  • Cas Number : 1269151-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73143
Availability details for
Quantity
  • Availability
  • Add to Cart