ENA313839068 Enamine

5,6-dimethyl-1-(prop-2-yn-1-yl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD18380716
  • Cas Number : 1269151-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73176
Availability details for
Quantity
  • Availability
  • Add to Cart