ENA313839075 Enamine

6-(piperazine-1-sulfonyl)-3,4-dihydro-2H-1,4-benzoxazin-3-one hydrochloride

  • MDL number MFCD18380724
  • Cas Number : 1269151-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-73219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73219
Availability details for
Quantity
  • Availability
  • Add to Cart