ENA313839080 Enamine

N-(3-aminopropyl)-N-cyclopropylcyclobutanecarboxamide hydrochloride

  • MDL number MFCD18380729
  • Cas Number : 1269152-74-0
  • Molecular Weight : 232.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-73265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73265
Availability details for
Quantity
  • Availability
  • Add to Cart