ENA313839089 Enamine

1-(4-fluorobenzoyl)-1,4-diazepane hydrochloride

  • MDL number MFCD18380739
  • Cas Number : 1269152-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73377
Availability details for
Quantity
  • Availability
  • Add to Cart