ENA313839090 Enamine

3-amino-6,7-dichloro-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD18380740
  • Cas Number : 1269151-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73384
Availability details for
Quantity
  • Availability
  • Add to Cart