ENA313839091 Enamine

N-(1-tert-butyl-4,5,6,7-tetrahydro-1H-indazol-4-yl)-2-chloroacetamide hydrochloride

  • MDL number MFCD18380741
  • Cas Number : 1269152-50-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73385
Availability details for
Quantity
  • Availability
  • Add to Cart