ENA313839093 Enamine

tert-butyl N-(2-amino-2-cyclopropylpropyl)carbamate

  • MDL number MFCD18380743
  • Cas Number : 1269152-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73398
Availability details for
Quantity
  • Availability
  • Add to Cart