ENA313839094 Enamine

1-(aminomethyl)-N-(1-phenylethyl)cyclopentan-1-amine

  • MDL number MFCD18380744
  • Cas Number : 1269152-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73419
Availability details for
Quantity
  • Availability
  • Add to Cart