ENA313839098 Enamine

3-[4-(trifluoromethyl)phenyl]cyclobutan-1-amine hydrochloride

  • MDL number MFCD18380747
  • Cas Number : 1269151-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73459
Availability details for
Quantity
  • Availability
  • Add to Cart