ENA313839104 Enamine

8-fluoro-5-methyl-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD18380752
  • Cas Number : 1269152-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73530
Availability details for
Quantity
  • Availability
  • Add to Cart