ENA313839105 Enamine

1-[(1-aminopropan-2-yl)sulfanyl]-4-bromobenzene hydrochloride

  • MDL number MFCD18380753
  • Cas Number : 1269152-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-73531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-73531
Availability details for
Quantity
  • Availability
  • Add to Cart