ENA313839106 Enamine

6-chloro-8-fluoro-1,2,3,4-tetrahydroquinoline hydrochloride

  • MDL number MFCD18380754
  • Cas Number : 1269152-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73538
Availability details for
Quantity
  • Availability
  • Add to Cart