ENA313839108 Enamine

5-(2-aminoethyl)-N,N,4-trimethyl-1,3-thiazol-2-amine

  • MDL number MFCD18380756
  • Cas Number : 1267179-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-73554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-73554
Availability details for
Quantity
  • Availability
  • Add to Cart