ENA313839141 Enamine

1-{1-[4-(propan-2-yl)phenyl]cyclopentyl}methanamine

  • MDL number MFCD11188968
  • Cas Number : 1037132-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72657
Availability details for
Quantity
  • Availability
  • Add to Cart