ENA313839150 Enamine

methyl 4-(4-bromobenzenesulfonyl)butanoate

  • MDL number MFCD12410101
  • Cas Number : 163496-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72668
Availability details for
Quantity
  • Availability
  • Add to Cart