ENA313839153 Enamine

2-hydroxy-2-(3-methoxyphenyl)acetic acid

  • MDL number MFCD00004229
  • Cas Number : 21150-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-65
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72676
Availability details for
Quantity
  • Availability
  • Add to Cart