ENA313839159 Enamine

[4-(4-bromophenoxy)-3-fluorophenyl]methanol

  • MDL number MFCD11190401
  • Cas Number : 1037141-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72687
Availability details for
Quantity
  • Availability
  • Add to Cart