ENA313839164 Enamine

4-[4-(2-methoxyethyl)phenoxy]aniline

  • MDL number MFCD11549248
  • Cas Number : 1099130-23-0
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72693
Availability details for
Quantity
  • Availability
  • Add to Cart