ENA313839166 Enamine

3-amino-4-phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD13704717
  • Cas Number : 1238343-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72695

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72695
Availability details for
Quantity
  • Availability
  • Add to Cart