ENA313839171 Enamine

4-chloro-2-[1-(propan-2-yl)-1H-pyrrole-2-amido]benzoic acid

  • MDL number MFCD13748324
  • Cas Number : 1155104-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72703

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72703
Availability details for
Quantity
  • Availability
  • Add to Cart