ENA313839174 Enamine

3-[(1-benzothiophen-3-yl)carbonyl]pyridine

  • MDL number MFCD11211448
  • Cas Number : 1094639-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72706

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72706
Availability details for
Quantity
  • Availability
  • Add to Cart