ENA313839178 Enamine

N-methyl-2-(methylamino)-N-(2-phenylethyl)benzamide

  • MDL number MFCD18380643
  • Cas Number : 173867-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72710
Availability details for
Quantity
  • Availability
  • Add to Cart