ENA313839184 Enamine

ethyl 2-[4-(hydroxymethyl)piperidin-1-yl]acetate

  • MDL number MFCD11133058
  • Cas Number : 1094301-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72717

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72717
Availability details for
Quantity
  • Availability
  • Add to Cart