ENA313839189 Enamine

1-butylpiperidin-4-amine

  • MDL number MFCD09742202
  • Cas Number : 50534-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-58151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58151
Availability details for
Quantity
  • Availability
  • Add to Cart