ENA313839191 Enamine

1-[2-(2-methylphenoxy)ethyl]piperazine

  • MDL number MFCD03231412
  • Cas Number : 65489-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72727
Availability details for
Quantity
  • Availability
  • Add to Cart